Gene Transcripts
- Gene Symbol
- Cyp11a1
- Entrez Gene ID
- 13070
- Gene Name
- cytochrome P450, family 11, subfamily a, polypeptide 1
- Gene Aliases
- Cyp11a, Cypxia1, D9Ertd411e, P450scc, Scc, cscc
- Chromosome Location
- Chr. 9: 58015017 - 58027023 on Build GRCm38
- UniGene ID
- Mm.302865
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK042878.1 |
- |
|
662 |
|
AK137397.1 |
- |
|
635 |
|
AK163559.1 |
- |
|
674 |
|
View Full Product Details