Gene Transcripts
- Gene Symbol
- Flg
- Entrez Gene ID
- 14246
- Gene Name
- filaggrin
- Gene Aliases
- AW107830, ft
- Chromosome Location
- Chr. 3: 93273546 - 93293689 on Build GRCm38
- UniGene ID
- Mm.162684
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK076261.1 |
- |
2 |
1124 |
|
AV230893.2 |
- |
1 |
492 |
|
J03458.1 |
- |
3 |
1322 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AA645085.1 |
- |
1 |
34 |
|
Chr. 3 - 93293487 - 93293693 [+] |
GenBankmRNA |
CJ238237.1 |
- |
1 |
303 |
|
Chr. 3 - 93293218 - 93293692 [+] |
GenBankmRNA |
BB371494.1 |
- |
1 |
119 |
|
Chr. 3 - 93293402 - 93293692 [+] |
GenBankmRNA |
CJ238937.1 |
- |
1 |
530 |
|
Chr. 3 - 93292991 - 93293691 [+] |
GenBankmRNA |
AA645130.1 |
- |
1 |
34 |
|
Chr. 3 - 93293487 - 93293693 [+] |
View Full Product Details