Gene Transcripts
- Gene Symbol
- TTTY22
- Entrez Gene ID
- 252954
- Gene Name
- testis-specific transcript, Y-linked 22 (non-protein coding)
- Gene Aliases
- NCRNA00147
- Chromosome Location
- Chr. Y: 9801153 - 9813245 on Build GRCh38
- UniGene ID
- Hs.511961
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001539.1 |
|
3 |
325 |
|
GenBank mRNA |
AF527834.1 |
- |
3 |
325 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AF527834.3 |
- |
3 |
325 |
|
Chr. Y - 9801153 - 9813245 [+] |
GENCODE |
ENST00000445253.1 |
- |
3 |
325 |
|
Chr. Y - 9801153 - 9813245 [+] |
View Full Product Details