Gene Transcripts
- Gene Symbol
- LRRTM3
- Entrez Gene ID
- 347731
- Gene Name
- leucine rich repeat transmembrane neuronal 3
- Gene Aliases
- Chromosome Location
- Chr. 10: 66926034 - 67101551 on Build GRCh38
- UniGene ID
- Hs.652155
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
DB029785.1 |
- |
5 |
520 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000422963.1 |
- |
2 |
186 |
|
Chr. 10 - 67052609 - 67055028 [+] |
View Full Product Details