Gene Transcripts
- Gene Symbol
- NHEG1
- Entrez Gene ID
- 100294720
- Gene Name
- neuroblastoma highly expressed 1
- Gene Aliases
- Chromosome Location
- Chr. 6: 136982159 - 136993231 on Build GRCh38
- UniGene ID
- Hs.520412
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027994.1 |
|
2 |
135 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000432330.5 |
- |
2 |
138 |
|
Chr. 6 - 136982165 - 136993234 [-] |
GENCODE |
ENST00000418699.1 |
- |
2 |
138 |
|
Chr. 6 - 136982165 - 136993234 [-] |
View Full Product Details