Gene Transcripts
- Gene Symbol
- GS1-594A7.3
- Entrez Gene ID
- 104798195
- Gene Name
- uncharacterized LOC104798195
- Gene Aliases
- Chromosome Location
- Chr. X: 15602881 - 15621484 on Build GRCh38
- UniGene ID
- Hs.131674
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_126564.1 |
|
2 |
277 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000421585.1 |
- |
2 |
277 |
|
Chr. X - 15602881 - 15621484 [+] |
View Full Product Details