Gene Transcripts

Gene Symbol
LINC01108
Entrez Gene ID
102216342
Gene Name
long intergenic non-protein coding RNA 1108
Gene Aliases
LncRNA-ES1
Chromosome Location
Chr. 6: 14280127 - 14285454 on Build GRCh38
UniGene ID
Hs.563409
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_108096.1 3 1875
NR_108097.1 2 1639
GenBank mRNA AK056826.1 - 2 1330


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000427276.1 - 2 1330 Chr. 6 - 14280127 - 14285145 [-]

View Full Product Details