Gene Transcripts
- Gene Symbol
- BSN-AS2
- Entrez Gene ID
- 100132677
- Gene Name
- BSN antisense RNA 2 (head to head)
- Gene Aliases
- Chromosome Location
- Chr. 3: 49549306 - 49554366 on Build GRCh38
- UniGene ID
- Hs.435651
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_038866.1 |
|
3 |
1978 |
|
GenBank mRNA |
AK096387.1 |
- |
3 |
1978 |
|
AK097709.1 |
- |
4 |
1818 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000421598.1 |
- |
3 |
1978 |
|
Chr. 3 - 49549306 - 49554366 [-] |
View Full Product Details