Gene Transcripts
- Gene Symbol
- SOX21-AS1
- Entrez Gene ID
- 100507533
- Gene Name
- SOX21 antisense RNA 1 (head to head)
- Gene Aliases
- Chromosome Location
- Chr. 13: 94712716 - 94715945 on Build GRCh38
- UniGene ID
- Hs.556071
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046514.1 |
|
2 |
2032 |
|
GenBank mRNA |
AK055459.1 |
- |
2 |
2032 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000438290.2 |
- |
2 |
2032 |
|
Chr. 13 - 94712716 - 94716246 [+] |
View Full Product Details