Gene Transcripts
- Gene Symbol
- SCNN1D
- Entrez Gene ID
- 6339
- Gene Name
- sodium channel epithelial 1 delta subunit
- Gene Aliases
- ENaCd, ENaCdelta, SCNED, dNaCh
- Chromosome Location
- Chr. 1: 1280415 - 1292029 on Build GRCh38
- UniGene ID
- Hs.512681
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK131558.1 |
- |
1 |
294 |
|
BX424308.2 |
- |
2 |
353 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000453732.1 |
- |
2 |
351 |
|
Chr. 1 - 1275237 - 1279393 [+] |
View Full Product Details