Gene Transcripts
- Gene Symbol
- DLEU2
- Entrez Gene ID
- 8847
- Gene Name
- deleted in lymphocytic leukemia 2 (non-protein coding)
- Gene Aliases
- 1B4, BCMSUN, DLB2, LEU2, LINC00022, MIR15AHG, NCRNA00022, RFP2OS, TRIM13OS
- Chromosome Location
- Chr. 13: 49982552 - 50125541 on Build GRCh38
- UniGene ID
- Hs.547964
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC038090.1 |
- |
1, 2 |
167 |
|
BC070292.1 |
- |
1, 2 |
174 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000421758.5 |
- |
1, 2 |
154 |
|
Chr. 13 - 50049189 - 50081973 [-] |
GENCODE |
ENST00000433070.6 |
- |
1, 2 |
173 |
|
Chr. 13 - 50044387 - 50081992 [-] |
View Full Product Details