Gene Transcripts
- Gene Symbol
- TRAF3IP2-AS1
- Entrez Gene ID
- 643749
- Gene Name
- TRAF3IP2 antisense RNA 1
- Gene Aliases
- C6UAS, C6orf3, NCRNA00248, TRAF3IP2-AS2
- Chromosome Location
- Chr. 6: 111483472 - 111602295 on Build GRCh38
- UniGene ID
- Hs.486228
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_034111.1 |
|
4 |
586 |
|
GenBank mRNA |
BC031311.1 |
- |
3 |
852 |
|
BX647114.1 |
- |
4 |
542 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000525151.5 |
- |
4 |
542 |
|
Chr. 6 - 111483516 - 111503608 [+] |
GENCODE |
ENST00000440001.2 |
- |
3 |
845 |
|
Chr. 6 - 111483665 - 111503611 [+] |
GenBankmRNA |
DB550044.1 |
- |
3 |
416 |
|
Chr. 6 - 111484094 - 111503610 [+] |
View Full Product Details