Gene Transcripts
- Gene Symbol
- CECR3
- Entrez Gene ID
- 27442
- Gene Name
- cat eye syndrome chromosome region, candidate 3 (non-protein coding)
- Gene Aliases
- Chromosome Location
- Chr. 22: 17256859 - 17266733 on Build GRCh38
- UniGene ID
- Hs.517394
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_038398.2 |
|
3 |
1825 |
|
GenBank mRNA |
AF277398.1 |
- |
3 |
1827 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000428828.1 |
- |
3 |
1825 |
|
Chr. 22 - 17256859 - 17266733 [-] |
View Full Product Details