Gene Transcripts
- Gene Symbol
- ZFAT-AS1
- Entrez Gene ID
- 594840
- Gene Name
- ZFAT antisense RNA 1
- Gene Aliases
- NCRNA00070, SAS-ZFAT, ZFAT-AS, ZFATAS
- Chromosome Location
- Chr. 8: 134598071 - 134600689 on Build GRCh38
- UniGene ID
- Hs.626298
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002438.1 |
|
1, 2 |
515 |
|
GenBank mRNA |
AB167742.1 |
- |
1, 2 |
515 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000505776.1 |
- |
1, 2 |
515 |
|
Chr. 8 - 134598071 - 134600689 [+] |
View Full Product Details