Gene Transcripts
- Gene Symbol
- NKX2-1-AS1
- Entrez Gene ID
- 100506237
- Gene Name
- NKX2-1 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 14: 36519278 - 36523016 on Build GRCh38
- UniGene ID
- Hs.497090
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_103710.1 |
|
1 |
207 |
|
GenBank mRNA |
BX161496.1 |
- |
1 |
169 |
|
DA256249.1 |
- |
1 |
207 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX383579.2 |
- |
1 |
166 |
|
Chr. 14 - 36519316 - 36521759 [+] |
GenBankmRNA |
BX367709.2 |
- |
1 |
166 |
|
Chr. 14 - 36519316 - 36522225 [+] |
GENCODE |
ENST00000521292.2 |
- |
1 |
207 |
|
Chr. 14 - 36519278 - 36523016 [+] |
View Full Product Details