Gene Transcripts
- Gene Symbol
- MIR9-3HG
- Entrez Gene ID
- 254559
- Gene Name
- MIR9-3 host gene
- Gene Aliases
- LINC00925
- Chromosome Location
- Chr. 15: 89361579 - 89398487 on Build GRCh38
- UniGene ID
- Hs.136313
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK123792.1 |
- |
1 |
102 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000546186.2 |
- |
1 |
102 |
|
Chr. 15 - 89386596 - 89396240 [+] |
View Full Product Details