Gene Transcripts
- Gene Symbol
- Timd2
- Entrez Gene ID
- 171284
- Gene Name
- T cell immunoglobulin and mucin domain containing 2
- Gene Aliases
- AA816106, AU019358, AU019662, C78111, G9D3, TIM-2, Tim2
- Chromosome Location
- Chr. 11: 46668960 - 46707104 on Build GRCm38
- UniGene ID
- Mm.234654
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001161355.1 |
NP_001154827.1
|
6, 7 |
941 |
|
NM_001161356.1 |
NP_001154828.1
|
6, 7 |
714 |
|
NM_134249.5 |
NP_599010.4
|
6, 7 |
823 |
|
GenBank mRNA |
AB009015.1 |
- |
6, 7 |
714 |
|
AF399827.1 |
- |
6, 7 |
675 |
|
AF399828.1 |
- |
6, 7 |
675 |
|
AK145630.1 |
- |
6, 7 |
702 |
|
AK163225.1 |
- |
6, 7 |
702 |
|
AK166877.1 |
- |
6, 7 |
695 |
|
BC025096.1 |
- |
6, 7 |
753 |
4158605 |
BC028829.1 |
- |
6, 7 |
785 |
5102129 |
View Full Product Details