Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081107.1 | NP_001074576.1 | 4 | 1344 | |
GenBank mRNA | AF436846.1 | - | 2 | 326 | |
AK050769.1 | - | 3 | 1142 | ||
AK052427.1 | - | 4 | 1304 | ||
BC082601.1 | - | 4 | 1308 | 30662103 | |
BC109169.2 | - | 4 | 1212 | ||
BC109170.2 | - | 4 | 1217 | 40048258 |