Gene Transcripts
- Gene Symbol
- Tmc2
- Entrez Gene ID
- 192140
- Gene Name
- transmembrane channel-like gene family 2
- Gene Aliases
- CWEA2
- Chromosome Location
- Chr. 2: 130195194 - 130264445 on Build GRCm38
- UniGene ID
- Mm.219546
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138655.1 |
NP_619596.1
|
12 |
1754 |
|
GenBank mRNA |
AF417581.1 |
- |
12 |
1754 |
|
View Full Product Details