Gene Transcripts

Gene Symbol
Gm5
Entrez Gene ID
194292
Gene Name
predicted gene 5
Gene Aliases
Gm1812
Chromosome Location
Chr. 5: 150499747 - 150502219 on Build GRCm38
UniGene ID
Mm.300184
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AA414041.1 - 1 76
CO797249.1 - 1 293

View Full Product Details