Gene Transcripts

Gene Symbol
Fchsd2
Entrez Gene ID
207278
Gene Name
FCH and double SH3 domains 2
Gene Aliases
BC034086, R74866, Sh3md3, mKIAA0769
Chromosome Location
Chr. 7: 101108711 - 101284405 on Build GRCm38
UniGene ID
Mm.263041
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146010.1 NP_001139482.1 8 918
NM_199012.2 NP_950177.2 8 918
GenBank mRNA AK122368.1 - 8 810
AK140322.1 - 8 897
AK141949.1 - 8 918
AK156841.1 - 7 744
AK157054.1 - 8 891
BC016532.1 - 8 917
BC060647.1 - 8 880 6854041

View Full Product Details