Gene Transcripts

Gene Symbol
Setd1b
Entrez Gene ID
208043
Gene Name
SET domain containing 1B
Gene Aliases
AA516740, BC035291, KMT2G, mKIAA1076
Chromosome Location
Chr. 5: 123140787 - 123168634 on Build GRCm38
UniGene ID
Mm.250391
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040398.2 NP_001035488.2 17 6178
GenBank mRNA BC023450.1 - 3 359
BC035291.1 - 9 2680
BC038367.1 - 11 3256
BC040775.1 - 9 2680
BC041681.1 - 9 2699

View Full Product Details