Gene Transcripts

Gene Symbol
Vmn2r15
Entrez Gene ID
211223
Gene Name
vomeronasal 2, receptor 15
Gene Aliases
EG211223
Chromosome Location
Chr. 5: 109286269 - 109297556 on Build GRCm38
UniGene ID
Mm.469881
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104626.1 NP_001098096.1 2 311

View Full Product Details