Gene Transcripts

Gene Symbol
Megf11
Entrez Gene ID
214058
Gene Name
multiple EGF-like-domains 11
Gene Aliases
2410080H04Rik, D130061K05
Chromosome Location
Chr. 9: 64385618 - 64709205 on Build GRCm38
UniGene ID
Mm.127721
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134399.1 NP_001127871.1 22 3029
NM_172522.4 NP_766110.3 21 2936
GenBank mRNA AK051642.1 - 21 2873
AK122555.2 - 23 3098

View Full Product Details