Gene Transcripts

Gene Symbol
Tph2
Entrez Gene ID
216343
Gene Name
tryptophan hydroxylase 2
Gene Aliases
AU043594, Ntph
Chromosome Location
Chr. 10: 115078641 - 115185022 on Build GRCm38
UniGene ID
Mm.31597
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173391.3 NP_775567.2 3 524
GenBank mRNA AY090565.1 - 3 524
BC120514.1 - 3 515 40129437
BC125430.1 - 3 515 40129945

View Full Product Details