Gene Transcripts

Gene Symbol
Ube2o
Entrez Gene ID
217342
Gene Name
ubiquitin-conjugating enzyme E2O
Gene Aliases
9630022H21, B230113M03Rik, E2-230K, mKIAA1734
Chromosome Location
Chr. 11: 116537740 - 116581476 on Build GRCm38
UniGene ID
Mm.243950
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173755.3 NP_776116.2 9 1326
GenBank mRNA AK129431.1 - 9 1326
BC059193.1 - 9 951 4160667
U20780.1 - 9 987

View Full Product Details