Gene Transcripts
- Gene Symbol
- Pomt2
- Entrez Gene ID
- 217734
- Gene Name
- protein-O-mannosyltransferase 2
- Gene Aliases
- A830009D15Rik, AW046274
- Chromosome Location
- Chr. 12: 87106861 - 87147902 on Build GRCm38
- UniGene ID
- Mm.388249
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_153415.3 |
NP_700464.2
|
19, 20 |
2514 |
|
GenBank mRNA |
AF246235.1 |
- |
19, 20 |
2118 |
|
AK043573.1 |
- |
19, 20 |
2215 |
|
AK043713.1 |
- |
19, 20 |
2059 |
|
AK050915.1 |
- |
19, 20 |
2083 |
|
AK076813.1 |
- |
19, 20 |
2517 |
|
AY090481.1 |
- |
19, 20 |
2514 |
|
AY090482.1 |
- |
19, 20 |
2114 |
|
BC052045.1 |
- |
18, 19 |
2167 |
6402328 |
BC062965.1 |
- |
18, 19 |
2167 |
|
BC079589.1 |
- |
19, 20 |
2084 |
|
View Full Product Details