Gene Transcripts

Gene Symbol
Gcnt4
Entrez Gene ID
218476
Gene Name
glucosaminyl (N-acetyl) transferase 4, core 2 (beta-1,6-N-acetylglucosaminyltransferase)
Gene Aliases
C2gnt3, Gm279, Gm73
Chromosome Location
Chr. 13: 96924352 - 96950914 on Build GRCm38
UniGene ID
Mm.483490
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166065.1 NP_001159537.1 3 1001
GenBank mRNA AK157788.1 - 3 1002

View Full Product Details