Gene Transcripts

Gene Symbol
March6
Entrez Gene ID
223455
Gene Name
membrane-associated ring finger (C3HC4) 6
Gene Aliases
3830408G03, F830029L24Rik, mKIAA0597
Chromosome Location
Chr. 15: 31455894 - 31531048 on Build GRCm38
UniGene ID
Mm.272185
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172606.2 NP_766194.2 12 1228
GenBank mRNA AK089827.1 - 12 1230
AK132461.1 - 12 1201
CF540592.1 - 6 290

View Full Product Details