Gene Transcripts

Gene Symbol
Mroh2b
Entrez Gene ID
223825
Gene Name
maestro heat-like repeat family member 2B
Gene Aliases
4930455B06Rik, Heatr7b2, SPIF
Chromosome Location
Chr. 15: 4898234 - 4962207 on Build GRCm38
UniGene ID
Mm.159281
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166066.1 NP_001159538.1 25, 26 3016
GenBank mRNA KF778378.1 - 24, 25 2596
KF778379.1 - 4, 5 616

View Full Product Details