Gene Transcripts

Gene Symbol
Dbx2
Entrez Gene ID
223843
Gene Name
developing brain homeobox 2
Gene Aliases
9430056A22Rik, 9630005K15, AI846217, Gm1229
Chromosome Location
Chr. 15: 95623563 - 95654771 on Build GRCm38
UniGene ID
Mm.152937
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207533.2 NP_997416.2 3 758
GenBank mRNA AK035803.1 - 3 355

View Full Product Details