Gene Transcripts
- Gene Symbol
- Thoc1
- Entrez Gene ID
- 225160
- Gene Name
- THO complex 1
- Gene Aliases
- 3110002N20Rik, AW107452, NMP-84
- Chromosome Location
- Chr. 18: 9958180 - 9995484 on Build GRCm38
- UniGene ID
- Mm.219648
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_153552.3 |
NP_705780.1
|
5, 6 |
382 |
|
GenBank mRNA |
AK031785.1 |
- |
5, 6 |
383 |
|
AK032200.1 |
- |
5, 6 |
383 |
|
AK042867.1 |
- |
5, 6 |
388 |
|
AK047991.1 |
- |
1, 2 |
1645 |
|
AK052835.1 |
- |
5, 6 |
378 |
|
AK139307.1 |
- |
5, 6 |
388 |
|
AK167374.1 |
- |
5, 6 |
396 |
|
AK168619.1 |
- |
5, 6 |
647 |
|
AK207368.1 |
- |
2, 3 |
157 |
|
AK215140.1 |
- |
2, 3 |
157 |
|
AK215219.1 |
- |
2, 3 |
157 |
|
BC024951.1 |
- |
5, 6 |
554 |
4010605 |
View Full Product Details