Gene Transcripts

Gene Symbol
Lzts2
Entrez Gene ID
226154
Gene Name
leucine zipper, putative tumor suppressor 2
Gene Aliases
BC014695, LAPSER1, mKIAA1813
Chromosome Location
Chr. 19: 45015176 - 45027115 on Build GRCm38
UniGene ID
Mm.28204
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130525.1 NP_001123997.1 5 1930
NM_001130526.1 NP_001123998.1 5 1922
NM_145503.2 NP_663478.2 5 1974
GenBank mRNA AK081367.1 - 5 1849
AK129446.1 - 4 1839
AK144154.1 - 5 1941
AK169936.1 - 5 1965
BC014695.1 - 4 1823 3157493
BC092263.1 - 5 1954 2654420

View Full Product Details