Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175467.3 | NP_780676.1 | 13 | 1785 | |
GenBank mRNA | AK048374.1 | - | 12 | 1646 | |
AK054240.1 | - | 12 | 1872 | ||
AK078679.1 | - | 12 | 1849 | ||
AK078686.1 | - | 13 | 1786 | ||
BC094496.1 | - | 12 | 1630 | 3483569 |