Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175467.3 | NP_780676.1 | 9 | 1289 | |
GenBank mRNA | AK048374.1 | - | 8 | 1150 | |
AK054240.1 | - | 8 | 1376 | ||
AK078679.1 | - | 8 | 1353 | ||
AK078686.1 | - | 9 | 1290 | ||
BC094496.1 | - | 8 | 1135 | 3483569 |