Gene Transcripts
- Gene Symbol
- Lppr4
- Entrez Gene ID
- 229791
- Gene Name
- lipid phosphate phosphatase-related protein type 4
- Gene Aliases
- A330086D10, D3Bwg0562e, PRG-1, mKIAA0455
- Chromosome Location
- Chr. 3: 117319145 - 117360876 on Build GRCm38
- UniGene ID
- Mm.140138
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_177664.5 |
NP_808332.3
|
6, 7 |
1607 |
|
GenBank mRNA |
AF541279.1 |
- |
6, 7 |
1608 |
|
AK046782.1 |
- |
6, 7 |
1608 |
|
AK079635.1 |
- |
6, 7 |
941 |
|
AK129149.1 |
- |
6, 7 |
1203 |
|
AY266266.1 |
- |
6, 7 |
1608 |
|
AY266267.1 |
- |
6, 7 |
1608 |
|
BC082323.1 |
- |
5, 6 |
2585 |
|
BC137701.1 |
- |
6, 7 |
1202 |
8860720 |
BC137702.1 |
- |
6, 7 |
1203 |
8860721 |
View Full Product Details