Gene Transcripts

Gene Symbol
Zfyve9
Entrez Gene ID
230597
Gene Name
zinc finger, FYVE domain containing 9
Gene Aliases
E030027L17, Madhip, NSP, SARA, SMADIP
Chromosome Location
Chr. 4: 108637834 - 108781148 on Build GRCm38
UniGene ID
Mm.196776
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183300.2 NP_899123.2 7 820
GenBank mRNA DQ303413.1 - 7 820

View Full Product Details