Gene Transcripts

Gene Symbol
Dok7
Entrez Gene ID
231134
Gene Name
docking protein 7
Gene Aliases
A930013K19Rik, AW049091, Dok-7, EF-12, Oit5
Chromosome Location
Chr. 5: 35056566 - 35087839 on Build GRCm38
UniGene ID
Mm.205483
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172708.3 NP_766296.1 3 298
GenBank mRNA AB220919.1 - 4 344
AK044445.1 - 3 299
BC089590.1 - 4 417 6759981

View Full Product Details