Gene Transcripts

Gene Symbol
Fbxl18
Entrez Gene ID
231863
Gene Name
F-box and leucine-rich repeat protein 18
Gene Aliases
AI427077, B130019G13Rik, C330021B20Rik
Chromosome Location
Chr. 5: 142871787 - 142895643 on Build GRCm38
UniGene ID
Mm.361645
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033312.3 NP_001028484.2 5 2373
GenBank mRNA AK049302.1 - 5 2327
AK144757.1 - 5 2375
AK154354.1 - 5 2353

View Full Product Details