Gene Transcripts

Gene Symbol
Ppp1r12c
Entrez Gene ID
232807
Gene Name
protein phosphatase 1, regulatory (inhibitor) subunit 12C
Gene Aliases
2410197A17Rik, AI839747, B430101M08, Mbs85
Chromosome Location
Chr. 7: 4481519 - 4502222 on Build GRCm38
UniGene ID
Mm.360391
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029834.3 NP_084110.2 18 2090
GenBank mRNA AK046570.1 - 18 2061
AK144691.1 - 18 2091
BC043678.1 - 4 267
BC114396.1 - 8 607

View Full Product Details