Gene Transcripts

Gene Symbol
Synm
Entrez Gene ID
233335
Gene Name
synemin, intermediate filament protein
Gene Aliases
4930412K21Rik, AI852401, Dmn, E130104F11, Syn, Synemin
Chromosome Location
Chr. 7: 67730161 - 67759742 on Build GRCm38
UniGene ID
Mm.60526
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183312.3 NP_899135.3 3 1077
NM_201639.2 NP_964001.2 3 1077
NM_207663.3 NP_997546.2 3 1077
GenBank mRNA AJ579700.1 - 3 1028
AJ579701.1 - 3 1028
AJ579702.1 - 3 1028
AK053514.1 - 2 95
AK143019.1 - 3 1078
AK172937.1 - 2 158
BC026872.1 - 4 921
BC040093.1 - 4 921

View Full Product Details