Gene Transcripts

Gene Symbol
Mfrp
Entrez Gene ID
259172
Gene Name
membrane-type frizzled-related protein
Gene Aliases
rd6
Chromosome Location
Chr. 9: 44101770 - 44109187 on Build GRCm38
UniGene ID
Mm.227110
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190314.1 NP_001177243.1 14 3827
NM_147126.3 NP_667337.1 14 3845

Gene Symbol
C1qtnf5
Entrez Gene ID
235312
Gene Name
C1q and tumor necrosis factor related protein 5
Gene Aliases
Ctrp5
Chromosome Location
Chr. 9: 44101770 - 44109187 on Build GRCm38
UniGene ID
Mm.227110
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040631.2 NP_001035721.1 3 829
NM_001040632.2 NP_001035722.1 3 920
NM_001190313.1 NP_001177242.1 14 3827
NM_001190319.1 NP_001177248.1 3 904
NM_145613.4 NP_663588.2 14 3845
GenBank mRNA AF469650.1 - 14 3773
AK053629.1 - 14 3829
BC023068.1 - 3 829 5355789
BC025174.1 - 3 788 3978387
BE861753.1 - 1 289
CA451902.1 - 1 163
DQ002398.1 - 3 828

View Full Product Details