Gene Transcripts

Gene Symbol
Lingo1
Entrez Gene ID
235402
Gene Name
leucine rich repeat and Ig domain containing 1
Gene Aliases
4930471K13Rik, AV148400, LERN1, LINGO-1, Lrrn6a, UNQ201
Chromosome Location
Chr. 9: 56618460 - 56796784 on Build GRCm38
UniGene ID
Mm.246605
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181074.4 NP_851419.1 4 1175
GenBank mRNA AF373780.1 - 1 727
AK027262.2 - 2 798
AK034084.1 - 2 1275
BC052384.1 - 4 1175 5703857
BC065696.1 - 4 1120 5685897

View Full Product Details