Gene Transcripts

Gene Symbol
Dleu7
Entrez Gene ID
239133
Gene Name
deleted in lymphocytic leukemia, 7
Gene Aliases
BC038059
Chromosome Location
Chr. 14: 62276229 - 62292979 on Build GRCm38
UniGene ID
Mm.127330
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173419.2 NP_775595.1 2 583
GenBank mRNA AK082171.1 - 3 345
BC038059.1 - 2 576 4235022
CK618986.1 - 2 583

View Full Product Details