Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002705.4 | NP_002696.3 | 19, 20 | 2495 | |
XM_006720902.3 | XP_006720965.1 | 19, 20 | 2459 | ||
XM_017023374.1 | XP_016878863.1 | 19, 20 | 2507 | ||
XM_017023375.1 | XP_016878864.1 | 19, 20 | 2519 | ||
GenBank mRNA | AB011140.1 | - | 10, 11 | 1417 | |
AF001691.1 | - | 19, 20 | 2495 | ||
AF013717.1 | - | 19, 20 | 2416 | ||
AK295623.1 | - | 7, 8 | 936 | ||
BC114620.1 | - | 19, 20 | 2455 | 40070172 |