Gene Transcripts

Gene Symbol
Psmb3
Entrez Gene ID
26446
Gene Name
proteasome (prosome, macropain) subunit, beta type 3
Gene Aliases
AL033320, C10-II
Chromosome Location
Chr. 11: 97703434 - 97713500 on Build GRCm38
UniGene ID
Mm.21874
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011971.4 NP_036101.1 5, 6 620
GenBank mRNA AF060092.1 - 5, 6 582
AK002331.1 - 5, 6 621
AK010488.1 - 5, 6 601
AK153162.1 - 5, 6 653
BC014783.1 - 5, 6 587 3500033
BC098233.1 - 4, 5 582 1547611

View Full Product Details