Gene Transcripts

Gene Symbol
Dmrt3
Entrez Gene ID
240590
Gene Name
doublesex and mab-3 related transcription factor 3
Gene Aliases
Chromosome Location
Chr. 19: 25610247 - 25623921 on Build GRCm38
UniGene ID
Mm.62785
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177360.3 NP_796334.2 2 830
GenBank mRNA AF541936.1 - 2 830
BC052041.1 - 2 824 6404988

View Full Product Details