Gene Transcripts

Gene Symbol
Kcnv2
Entrez Gene ID
240595
Gene Name
potassium channel, subfamily V, member 2
Gene Aliases
KV11.1
Chromosome Location
Chr. 19: 27322060 - 27337179 on Build GRCm38
UniGene ID
Mm.269759
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183179.1 NP_899002.1 2 1775
GenBank mRNA AY238939.1 - 2 1643
BC039042.1 - 2 1775 5363843
BC046490.1 - 2 1806

View Full Product Details