Gene Transcripts

Gene Symbol
Slc35g1
Entrez Gene ID
240660
Gene Name
solute carrier family 35, member G1
Gene Aliases
AA590464, D330039I19Rik, Tmem20
Chromosome Location
Chr. 19: 38395980 - 38405607 on Build GRCm38
UniGene ID
Mm.262085
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175507.3 NP_780716.1 3 738
GenBank mRNA AK041618.1 - 3 738
AK052365.1 - 3 642
AK171987.1 - 3 670
AK209992.1 - 1 86
BC137663.1 - 3 749 8860682
BC137664.1 - 3 749 8860683

View Full Product Details