Gene Transcripts

Gene Symbol
Sbno1
Entrez Gene ID
243272
Gene Name
sno, strawberry notch homolog 1 (Drosophila)
Gene Aliases
9330180L10Rik, AI849128, AW557836, BC021875, Sbno, mSno1, sno
Chromosome Location
Chr. 5: 124368702 - 124426011 on Build GRCm38
UniGene ID
Mm.23879
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081203.1 NP_001074672.1 17 2305
GenBank mRNA AB188811.1 - 16 2234
AK077133.1 - 17 2311
AK133096.1 - 9 1129
BC021875.1 - 16 2251 4212192
BC138418.1 - 17 2362 8861438

View Full Product Details